missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromocrotonate, 85%, tech.

Catalog No. AC126450250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126450250 25 g Glass bottle
AC126451000 100 g Glass bottle
AC126455000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC126450250 Supplier Thermo Scientific Chemicals Supplier No. 126450250

CAS: 1117-71-1 | C5H7BrO2 | 179.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1117-71-1
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00000246,MFCD09998308
InChI Key RWIKCBHOVNDESJ-NSCUHMNNSA-N
Synonym methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester
PubChem CID 5369175
SMILES COC(=O)\C=C\CBr

Specifications

Color Yellow
Density 1.5200g/mL
Boiling Point 83.0°C to 85.0°C (13.0 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH=CHCO2CH3
Refractive Index 1.4990 to 1.5020
Quantity 25 g
Specific Gravity 1.52
IUPAC Name methyl (2E)-4-bromobut-2-enoate
Formula Weight 179.01
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl 4-bromocrotonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-251-

RUO – Research Use Only