missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Bromobenzoate 98.0+%, TCI America™
Supplier: TCI America B055625G
Specifications
| Methyl 4-Bromobenzoate | |
| 79°C | |
| C8H7BrO2 | |
| 25 g | |
| CZNGTXVOZOWWKM-UHFFFAOYSA-N | |
| methyl 4-bromobenzoate | |
| 12081 | |
| ≥98.0% (GC) |
| 619-42-1 | |
| White | |
| MFCD00013531 | |
| benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate | |
| COC(=O)C1=CC=C(C=C1)Br | |
| 215.046 | |
| 215.05 | |
| Crystalline Powder |
Chemical Identifiers
| 619-42-1 | |
| 215.046 | |
| CZNGTXVOZOWWKM-UHFFFAOYSA-N | |
| 12081 | |
| COC(=O)C1=CC=C(C=C1)Br |
| C8H7BrO2 | |
| MFCD00013531 | |
| benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate | |
| methyl 4-bromobenzoate |
Safety and Handling
TSCA : Yes