missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromo-3-hydroxybenzoate 98.0+%, TCI America™

Catalog No. M22465G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22465G 5 g
M22461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22465G Supplier TCI America Supplier No. M22465G

Chemical Identifiers

CAS 106291-80-9
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD08056372
InChI Key VYOFPLOREOHCDP-UHFFFAOYSA-N
Synonym 4-Bromo-3-hydroxybenzoic Acid Methyl Ester
PubChem CID 6425184
IUPAC Name methyl 4-bromo-3-hydroxybenzoate
SMILES COC(=O)C1=CC(O)=C(Br)C=C1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Bromo-3-hydroxybenzoate