missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Bromo-3-chlorobenzoate 96.0+%, TCI America™
$128.62 - $382.13
Chemical Identifiers
| CAS | 117738-74-6 |
|---|---|
| Molecular Formula | C8H6BrClO2 |
| Molecular Weight (g/mol) | 249.49 |
| MDL Number | MFCD11045619 |
| InChI Key | BPJMNAWSHVKGHE-UHFFFAOYSA-N |
| Synonym | 4-Bromo-3-chlorobenzoic Acid Methyl Ester |
| PubChem CID | 22366763 |
| IUPAC Name | methyl 4-bromo-3-chlorobenzoate |
| SMILES | COC(=O)C1=CC(Cl)=C(Br)C=C1 |
Chemical Identifiers
| 117738-74-6 | |
| 249.49 | |
| BPJMNAWSHVKGHE-UHFFFAOYSA-N | |
| 22366763 | |
| COC(=O)C1=CC(Cl)=C(Br)C=C1 |
| C8H6BrClO2 | |
| MFCD11045619 | |
| 4-Bromo-3-chlorobenzoic Acid Methyl Ester | |
| methyl 4-bromo-3-chlorobenzoate |
Specifications
| 117738-74-6 | |
| C8H6BrClO2 | |
| 1 g | |
| BPJMNAWSHVKGHE-UHFFFAOYSA-N | |
| methyl 4-bromo-3-chlorobenzoate | |
| 22366763 | |
| ≥96.0% (GC) | |
| Methyl 4-Bromo-3-chlorobenzoate |
| 59°C | |
| MFCD11045619 | |
| 4-Bromo-3-chlorobenzoic Acid Methyl Ester | |
| COC(=O)C1=CC(Cl)=C(Br)C=C1 | |
| 249.49 | |
| 249.49 | |
| Crystalline Powder |
Safety and Handling
TSCA : No