missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromo-2-methoxybenzoate 98.0+%, TCI America™

Catalog No. M25865G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25865G Supplier TCI America Supplier No. M25865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139102-34-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD06797867
InChI Key WPGAGRPPDYAZAD-UHFFFAOYSA-N
Synonym methyl4-bromo-2-methoxybenzoate, benzoic acid, 4-bromo-2-methoxy-, methyl ester, 4-bromo-2-methoxybenzoic acid methyl ester, ablock ab-14-0789, benzoic acid,4-bromo-2-methoxy-, methyl ester, pubchem16069, acmc-209ciq, methyl-4-bromo-2-methoxybenzoate, zerenex e/1100188, 4-bromo-2-methoxybenzoate 98
PubChem CID 15128242
IUPAC Name methyl 4-bromo-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1)Br)C(=O)OC

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 4-Bromo-2-methoxybenzoate