missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-2-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog No. AAH2777309
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2777309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2777309 Supplier Thermo Scientific Chemicals Supplier No. H2777309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 139102-34-4 | Molecular Formula: C9H9BrO3 | Molecular Weight: 245.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139102-34-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD06797867
InChI Key WPGAGRPPDYAZAD-UHFFFAOYSA-N
PubChem CID 15128242
IUPAC Name methyl 4-bromo-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1)Br)C(=O)OC

Specifications

Melting Point 32°C to 34°C
Boiling Point 100°C to 108°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Synonym methyl4-bromo-2-methoxybenzoate, benzoic acid, 4-bromo-2-methoxy-, methyl ester, 4-bromo-2-methoxybenzoic acid methyl ester, ablock ab-14-0789, benzoic acid,4-bromo-2-methoxy-, methyl ester, pubchem16069, acmc-209ciq, methyl-4-bromo-2-methoxybenzoate, zerenex e/1100188, 4-bromo-2-methoxybenzoate 98
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 4-bromo-2-methoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only