missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromo-2-chlorobenzoate 98.0+%, TCI America™

Catalog No. M28831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28831G 1 g
M28835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28831G Supplier TCI America Supplier No. M28831G

Chemical Identifiers

CAS 185312-82-7
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.488
MDL Number MFCD10566823
InChI Key JSNXIWYWUKTWAX-UHFFFAOYSA-N
Synonym 4-Bromo-2-chlorobenzoic Acid Methyl Ester
PubChem CID 22058730
IUPAC Name methyl 4-bromo-2-chlorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)Br)Cl

Specifications

Quantity 1 g
Formula Weight 249.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Bromo-2-chlorobenzoate