missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Aminosalicylate 98.0+%, TCI America™

Catalog No. M269925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M269925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M269925G Supplier TCI America Supplier No. M269925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4136-97-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00088091
InChI Key QQOXBFUTRLDXDP-UHFFFAOYSA-N
Synonym methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
PubChem CID 77787
ChEBI CHEBI:35089
IUPAC Name methyl 4-amino-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)N)O

Specifications

Melting Point 119°C
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Aminosalicylate
EINECSNumber (9)-2085
RTECSNumber VO1683000
TSCA No