missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminosalicylate, 97%, Thermo Scientific Chemicals

Catalog No. p-7059784
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4552022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4552022 Supplier Thermo Scientific Chemicals Supplier No. 04552022
Only null left

Methyl 4-aminosalicylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4136-97-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00088091
InChI Key QQOXBFUTRLDXDP-UHFFFAOYSA-N
Synonym methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
PubChem CID 77787
ChEBI CHEBI:35089
IUPAC Name methyl 4-amino-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)N)O

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 167.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 4-aminosalicylate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 223-958-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only