missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Aminopyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7133464
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28331G 1 g
M28335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28331G Supplier TCI America Supplier No. M28331G
Only null left

Chemical Identifiers

CAS 71469-93-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00956000
InChI Key YHOVYZINCVIRGK-UHFFFAOYSA-N
Synonym 4-Aminopyridine-2-carboxylic Acid Methyl Ester, Methyl 4-Aminopicolinate, 4-Aminopicolinic Acid Methyl Ester
PubChem CID 3759117
IUPAC Name methyl 4-aminopyridine-2-carboxylate
SMILES COC(=O)C1=NC=CC(=C1)N

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Aminopyridine-2-carboxylate