CAS RN 71469-93-7
IUPAC Name:
methyl 4-aminopyridine-2-carboxylate
Synonyms:
methyl 4-aminopyridine-2-carboxylate
Molecular Weight (g/mol):
152.15
Molecular Formula:
C7H8N2O2
InChi Key:
YHOVYZINCVIRGK-UHFFFAOYSA-N
SMILES:
COC(=O)C1=NC=CC(N)=C1
1
–
1
of
1
results
1
Methyl 4-Aminopyridine-2-carboxylate 98.0+%, TCI America™
CAS: 71469-93-7 Molecular Formula: C7H8N2O2 Molecular Weight (g/mol): 152.153 MDL Number: MFCD00956000 InChI Key: YHOVYZINCVIRGK-UHFFFAOYSA-N Synonym: 4-Aminopyridine-2-carboxylic Acid Methyl Ester, Methyl 4-Aminopicolinate, 4-Aminopicolinic Acid Methyl Ester PubChem CID: 3759117 IUPAC Name: methyl 4-aminopyridine-2-carboxylate SMILES: COC(=O)C1=NC=CC(=C1)N