missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(Aminomethyl)benzoate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133087
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22001G 1 g
M22005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22001G Supplier TCI America Supplier No. M22001G

Chemical Identifiers

CAS 6232-11-7
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00182671
InChI Key GIZCKBSSWNIUMZ-UHFFFAOYSA-N
Synonym methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride
PubChem CID 2729253
IUPAC Name methyl 4-(aminomethyl)benzoate;hydrochloride
SMILES COC(=O)C1=CC=C(C=C1)CN.Cl

Specifications

Melting Point 238°C
Color White
Quantity 1 g
Formula Weight 201.65
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-(Aminomethyl)benzoate Hydrochloride