Learn More
Methyl 4-(aminomethyl)benzoate hydrochloride, 97%
CAS: 6232-11-7 | C9H11NO2·ClH | 201.65 g/mol
$56.70 - $56.70
Chemical Identifiers
| CAS | 6232-11-7 |
|---|---|
| Molecular Formula | C9H11NO2·ClH |
| Molecular Weight (g/mol) | 201.65 |
| MDL Number | MFCD00182671 |
| InChI Key | GIZCKBSSWNIUMZ-UHFFFAOYSA-N |
| Synonym | methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride |
| PubChem CID | 2729253 |
| IUPAC Name | methyl 4-(aminomethyl)benzoate;hydrochloride |
| SMILES | COC(=O)C1=CC=C(C=C1)CN.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC396780010
|
Thermo Scientific Chemicals
396780010 |
1 g | Glass bottle |
Each for $56.70
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6232-11-7 | |
| 201.65 | |
| GIZCKBSSWNIUMZ-UHFFFAOYSA-N | |
| 2729253 | |
| COC(=O)C1=CC=C(C=C1)CN.Cl |
| C9H11NO2·ClH | |
| MFCD00182671 | |
| methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride | |
| methyl 4-(aminomethyl)benzoate;hydrochloride |
Specifications
| 6232-11-7 | |
| White | |
| 96.0 to 104% (Argentometry) | |
| C9H11NO2·ClH | |
| MFCD00182671 | |
| methyl 4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hydrochloride, methyl-4-aminomethyl benzoate hydrochloride, methyl 4-aminomethylbenzoate hcl, methyl 4-aminomethyl benzoate, hcl, 4-aminomethyl benzoic acid methyl ester hydrochloride, methyl4-aminomethyl benzoatehydrochloride, methyl 4-aminomethyl-benzoate hydrochloride, 4-aminomethyl benzoic acid methyl ester hcl, 4-aminomethylbenzoic acid methyl ester hydrochloride | |
| COC(=O)C1=CC=C(C=C1)CN.Cl | |
| 201.65 | |
| 201.65 | |
| Crystalline Powder |
| 234.5°C to 237.0°C | |
| Authentic | |
| Glass bottle | |
| NH2CH2C6H4COOCH3 | |
| 1 g | |
| GIZCKBSSWNIUMZ-UHFFFAOYSA-N | |
| methyl 4-(aminomethyl)benzoate;hydrochloride | |
| 2729253 | |
| 97% | |
| Methyl 4-(aminomethyl)benzoate hydrochloride |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only