missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Amino-3-methylbenzoate 98.0+%, TCI America™
$78.29 - $311.30
Chemical Identifiers
| CAS | 18595-14-7 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| MDL Number | MFCD00102230 |
| InChI Key | ZHIPSMIKSRYZFV-UHFFFAOYSA-N |
| Synonym | 4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate |
| PubChem CID | 2736799 |
| IUPAC Name | methyl 4-amino-3-methylbenzoate |
| SMILES | COC(=O)C1=CC=C(N)C(C)=C1 |
Chemical Identifiers
| 18595-14-7 | |
| 165.19 | |
| ZHIPSMIKSRYZFV-UHFFFAOYSA-N | |
| 2736799 | |
| COC(=O)C1=CC=C(N)C(C)=C1 |
| C9H11NO2 | |
| MFCD00102230 | |
| 4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate | |
| methyl 4-amino-3-methylbenzoate |
Specifications
| 18595-14-7 | |
| C9H11NO2 | |
| 5 g | |
| ZHIPSMIKSRYZFV-UHFFFAOYSA-N | |
| methyl 4-amino-3-methylbenzoate | |
| 2736799 | |
| ≥98.0% (GC,T) | |
| Methyl 4-Amino-3-methylbenzoate |
| 120°C | |
| MFCD00102230 | |
| 4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate | |
| COC(=O)C1=CC=C(N)C(C)=C1 | |
| 165.19 | |
| 165.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No