missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-methylbenzoate, 98%

Catalog No. AAH6111714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6111714 25 g
AAH6111722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6111714 Supplier Thermo Scientific Chemicals Supplier No. H6111714

CAS: 18595-14-7 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18595-14-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00102230
InChI Key ZHIPSMIKSRYZFV-UHFFFAOYSA-N
Synonym 4-amino-3-methylbenzoic acid methyl ester, 4-amino-3-toluic acid methyl ester, methyl 3-methyl-4-aminobenzoate, methyl-4-amino-3-methyl-benzoate, methyl 4-amino-3-methyl-benzoate, benzoic acid, 4-amino-3-methyl-, methyl ester, 4-amino-m-toluic acid methyl ester, methyl 4-amino-3-methylbenzenecarboxylate, zlchem 944, methyl 4-amino-m-toluate
PubChem CID 2736799
IUPAC Name methyl 4-amino-3-methylbenzoate
SMILES COC(=O)C1=CC=C(N)C(C)=C1

Specifications

Melting Point 116°C to 120°C
Quantity 25 g
Formula Weight 165.19
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-amino-3-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only