missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Amino-3-chlorobenzoate 97.0+%, TCI America™

Catalog No. p-7133375
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26981G 1 g
M26985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26981G Supplier TCI America Supplier No. M26981G
Only null left

Chemical Identifiers

CAS 84228-44-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD01320686
InChI Key WKVJBYKFGNVWLM-UHFFFAOYSA-N
Synonym 4-amino-3-chlorobenzoic acid methyl ester, 4-amino-3-chloro-benzoic acid methyl ester, methyl 4-amino-3-chloro-benzoate, benzoic acid, 4-amino-3-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl aniline, pubchem10804, akos bb-3098, timtec-bb sbb003712, acmc-20a51m, methyl-4-amino-3-chlorobenzoate
PubChem CID 1515284
IUPAC Name methyl 4-amino-3-chlorobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)N)Cl

Specifications

Melting Point 109°C
Quantity 1 g
Formula Weight 185.61
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Amino-3-chlorobenzoate