missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Amino-3-bromobenzoate 98.0+%, TCI America™

Catalog No. M20841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20841G 1 g
M20845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20841G Supplier TCI America Supplier No. M20841G

Chemical Identifiers

CAS 106896-49-5
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD01861385
InChI Key AIUWAOALZYWQBX-UHFFFAOYSA-N
Synonym 4-amino-3-bromo-benzoic acid methyl ester, methyl4-amino-3-bromobenzoate, 2-bromo-4-methoxycarbonylaniline, benzoic acid, 4-amino-3-bromo-, methyl ester, 4-amino-3-bromobenzoic acid methyl ester, acmc-2098mp, methyl-4-amino-3-bromobenzoate, methyl 4-amino-3-bromo-benzoate, timtec-bb sbb012847, 2-bromo-4-methoxycarbonyl aniline
PubChem CID 1515280
IUPAC Name methyl 4-amino-3-bromobenzoate
SMILES COC(=O)C1=CC=C(N)C(Br)=C1

Specifications

Melting Point 107°C
Quantity 1 g
Formula Weight 230.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Amino-3-bromobenzoate