missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-bromobenzoate, 97%

Catalog No. p-7055665
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6430303 1 g
AAH6430306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6430303 Supplier Thermo Scientific Chemicals Supplier No. H6430303
Only null left

CAS: 106896-49-5 | C8H8BrNO2 | 230.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106896-49-5
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD01861385
InChI Key AIUWAOALZYWQBX-UHFFFAOYSA-N
Synonym 4-amino-3-bromo-benzoic acid methyl ester, methyl4-amino-3-bromobenzoate, 2-bromo-4-methoxycarbonylaniline, benzoic acid, 4-amino-3-bromo-, methyl ester, 4-amino-3-bromobenzoic acid methyl ester, acmc-2098mp, methyl-4-amino-3-bromobenzoate, methyl 4-amino-3-bromo-benzoate, timtec-bb sbb012847, 2-bromo-4-methoxycarbonyl aniline
PubChem CID 1515280
IUPAC Name methyl 4-amino-3-bromobenzoate
SMILES COC(=O)C1=CC=C(N)C(Br)=C1

Specifications

Melting Point 105°C to 109°C
Quantity 1 g
Beilstein 4905039
Formula Weight 230.06
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 4-amino-3-bromobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only