missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-3-bromo-5-nitrobenzoate, 97%, Thermo Scientific Chemicals

Catalog No. AAH3321606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3321606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3321606 Supplier Thermo Scientific Chemicals Supplier No. H3321606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105655-17-2 | Molecular Formula: C8H7BrN2O4 | Molecular Weight: 275.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105655-17-2
Molecular Formula C8H7BrN2O4
Molecular Weight (g/mol) 275.058
MDL Number MFCD08443938
InChI Key PRTLMEWWYBJZPN-UHFFFAOYSA-N
Synonym methyl 4-amino-3-bromo-5-nitrobenzenecarboxylate, benzoic acid, 4-amino-3-bromo-5-nitro-, methyl ester, acmc-20aq1i, methylaminobromonitrobenzenecarboxylate, 3-nitro-4-amino-5-bromobenzoic acid methyl ester, benzoicacid,4-amino-3-bromo-5-nitro-,methylester, 4-amino-3-bromo-5-nitro-benzoic acid methyl ester
PubChem CID 13685347
IUPAC Name methyl 4-amino-3-bromo-5-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)Br)N)[N+](=O)[O-]

Specifications

Melting Point 107°C to 109°C
Quantity 5 g
Formula Weight 275.06
Percent Purity 97%
Chemical Name or Material Methyl 4-amino-3-bromo-5-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only