missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-2-methoxybenzoate, 98%, Thermo Scientific Chemicals

Catalog No. p-7021061
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1323906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1323906 Supplier Thermo Scientific Chemicals Supplier No. A1323906

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27492-84-8
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00017202
InChI Key YUPQMVSYNJQULF-UHFFFAOYSA-N
Synonym methyl 4-amino-o-anisate, 4-amino-2-methoxybenzoic acid methyl ester, benzoic acid, 4-amino-2-methoxy-, methyl ester, methyl 2-methoxy-4-aminobenzoate, methyl 4-amino-2-methoxy-benzoate, 4-amino-o-anisic acid methyl ester, methyl 4-amino-2-methoxybenzenecarboxylate, 2-methoxy-4-aminobenzoic acid methyl ester, ethylbenzyl dimethyltetradecylammonium chloride, pubchem4712
PubChem CID 168705
IUPAC Name methyl 4-amino-2-methoxybenzoate
SMILES COC(=O)C1=CC=C(N)C=C1OC

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Formula Weight 181.19
Percent Purity 98%
Chemical Name or Material Methyl 4-amino-2-methoxybenzoate
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 248-494-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only