missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Acetoxybenzoate 98.0+%, TCI America™

Catalog No. A122725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A122725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A122725G Supplier TCI America Supplier No. A122725G
Only null left

Chemical Identifiers

CAS 24262-66-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00017517
InChI Key PGJQPLVEUVHSFQ-UHFFFAOYSA-N
Synonym 4-Acetoxybenzoic Acid Methyl Ester
PubChem CID 578495
ChEBI CHEBI:86900
IUPAC Name methyl 4-acetyloxybenzoate
SMILES CC(=O)OC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 82°C
Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Acetoxybenzoate