missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-Dimethoxyanthranilate 98.0+%, TCI America™

Catalog No. D274925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D274925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D274925G Supplier TCI America Supplier No. D274925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26759-46-6
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.22
MDL Number MFCD00014904
InChI Key QQFHCCQSCQBKBG-UHFFFAOYSA-N
Synonym methyl 6-aminoveratrate, methyl 4,5-dimethoxyanthranilate, methyl 2-amino-4,5-dimethoxy benzoate, 2-amino-4,5-dimethoxybenzoic acid methyl ester, 6-aminoveratric acid methyl ester, methyl2-amino-4,5-dimethoxybenzoate, 4,5-dimethoxyanthranilic acid methyl, benzoic acid, 2-amino-4,5-dimethoxy-, methyl ester, methyl-2-amino-4,5-dimethoxybenzoate, 2-amino-4,5-dimethoxy-benzoic acid methyl ester
PubChem CID 611144
IUPAC Name methyl 2-amino-4,5-dimethoxybenzoate
SMILES COC(=O)C1=CC(OC)=C(OC)C=C1N

Specifications

Melting Point 129°C
Quantity 25 g
Formula Weight 211.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4,5-Dimethoxyanthranilate