missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate 98.0+%, TCI America™

Catalog No. M24265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24265G Supplier TCI America Supplier No. M24265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 155405-80-4
Molecular Formula C16H16N4O3
Molecular Weight (g/mol) 312.329
MDL Number MFCD17976681
InChI Key GDEKSBCRJKAARB-UHFFFAOYSA-N
Synonym 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid Methyl Ester, 2-Amino-4,7-dihydro-5-[2-[4-(methoxycarbonyl)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine
PubChem CID 11023369
IUPAC Name methyl 4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)CCC2=CNC3=C2C(=O)N=C(N3)N

Specifications

Quantity 5 g
Formula Weight 312.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate