accessibility menu, dialog, popup

UserName

CAS RN 155405-80-4

IUPAC Name: methyl 4-(2-{2-amino-4-oxo-1H,4H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)benzoate
Synonyms: methyl 4-(2-{2-amino-4-oxo-1H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)benzoate
Molecular Weight (g/mol): 312.33
Molecular Formula: C16H16N4O3
InChi Key: GDEKSBCRJKAARB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CCC2=CNC3=C2C(=O)N=C(N)N3)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results