missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(Trifluoromethyl)benzoate 98.0+%, TCI America™
$29.83 - $78.29
Chemical Identifiers
| CAS | 2557-13-3 |
|---|---|
| Molecular Formula | C9H7F3O2 |
| Molecular Weight (g/mol) | 204.148 |
| InChI Key | QQHNNQCWKYFNAC-UHFFFAOYSA-N |
| Synonym | methyl 3-trifluoromethyl benzoate, 3-carbomethoxybenzotrifluoride, 3-trifluoromethyl benzoic acid methyl ester, methyl3-trifluoromethyl benzoate, methyl m-trifluoromethyl benzoate, 3-trifluoromethyl-benzoic aic methyl ester, mtf-bom, methyl m-trifluoromethylbenzoate, 3-methoxycarbonyl benzotrifluoride |
| PubChem CID | 520213 |
| IUPAC Name | methyl 3-(trifluoromethyl)benzoate |
| SMILES | COC(=O)C1=CC(=CC=C1)C(F)(F)F |
Chemical Identifiers
| 2557-13-3 | |
| 204.148 | |
| methyl 3-trifluoromethyl benzoate, 3-carbomethoxybenzotrifluoride, 3-trifluoromethyl benzoic acid methyl ester, methyl3-trifluoromethyl benzoate, methyl m-trifluoromethyl benzoate, 3-trifluoromethyl-benzoic aic methyl ester, mtf-bom, methyl m-trifluoromethylbenzoate, 3-methoxycarbonyl benzotrifluoride | |
| methyl 3-(trifluoromethyl)benzoate |
| C9H7F3O2 | |
| QQHNNQCWKYFNAC-UHFFFAOYSA-N | |
| 520213 | |
| COC(=O)C1=CC(=CC=C1)C(F)(F)F |
Specifications
| 2557-13-3 | |
| 198°C | |
| 5 g | |
| methyl 3-trifluoromethyl benzoate, 3-carbomethoxybenzotrifluoride, 3-trifluoromethyl benzoic acid methyl ester, methyl3-trifluoromethyl benzoate, methyl m-trifluoromethyl benzoate, 3-trifluoromethyl-benzoic aic methyl ester, mtf-bom, methyl m-trifluoromethylbenzoate, 3-methoxycarbonyl benzotrifluoride | |
| COC(=O)C1=CC(=CC=C1)C(F)(F)F | |
| 204.148 | |
| 204.15 | |
| Liquid |
| Yellow | |
| C9H7F3O2 | |
| 3272 | |
| QQHNNQCWKYFNAC-UHFFFAOYSA-N | |
| methyl 3-(trifluoromethyl)benzoate | |
| 520213 | |
| ≥98.0% (GC) | |
| Methyl 3-(Trifluoromethyl)benzoate |
Safety and Handling
TSCA : No