missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(trifluoromethyl)benzoate, 97%

Catalog No. AAA1431606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1431606 5 g
AAA1431614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1431606 Supplier Thermo Scientific Chemicals Supplier No. A1431606

CAS: 2557-13-3 | C9H7F3O2 | 204.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2557-13-3
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00043543
InChI Key QQHNNQCWKYFNAC-UHFFFAOYSA-N
Synonym methyl 3-trifluoromethyl benzoate, 3-carbomethoxybenzotrifluoride, 3-trifluoromethyl benzoic acid methyl ester, methyl3-trifluoromethyl benzoate, methyl m-trifluoromethyl benzoate, 3-trifluoromethyl-benzoic aic methyl ester, mtf-bom, methyl m-trifluoromethylbenzoate, 3-methoxycarbonyl benzotrifluoride
PubChem CID 520213
IUPAC Name methyl 3-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Density 1.295
Boiling Point 80°C to 82°C (15 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.453
Quantity 5 g
Beilstein 1963428
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material Methyl 3-(trifluoromethyl)benzoate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only