missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Sulfolene-3-carboxylate 98.0+%, TCI America™

Catalog No. S04761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04761G 1 g
S04765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04761G Supplier TCI America Supplier No. S04761G

Chemical Identifiers

CAS 67488-50-0
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.21
MDL Number MFCD00042754
InChI Key FHYQDMHVFCUXLL-UHFFFAOYSA-N
Synonym methyl 2,5-dihydrothiophene-3-carboxylate 1,1-dioxide, 3-methoxycarbonyl-3-sulfolene, methyl 3-sulfolene-3-carboxylate, 3-methoxycarbonyl-3-sulpholene, 3-sulfolene-3-carboxylic acid methyl ester, 2,5-dihydro-3-methoxycarbonylthiophene 1,1-dioxide, 3-methoxycarbonyl-2,5-dihydrothiophene-1,1-dioxide, methyl 1,1-dioxo-2,5-dihydro-1??-thiophene-3-carboxylate, 3-carbomethoxysulpholene, 3-methoxycarbonyl-2,1-dioxide
PubChem CID 368979
IUPAC Name methyl 2-methyl-1,1-dioxo-4,5-dihydro-1λ⁶-thiophene-3-carboxylate
SMILES COC(=O)C1=C(C)S(=O)(=O)CC1

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 1 g
Formula Weight 176.19
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Sulfolene-3-carboxylate