accessibility menu, dialog, popup

UserName

CAS RN 67488-50-0

IUPAC Name: methyl 2-methyl-1,1-dioxo-4,5-dihydro-1λ⁶-thiophene-3-carboxylate
Synonyms: methyl 2-methyl-1,1-dioxo-4,5-dihydro-1λ⁶-thiophene-3-carboxylate
Molecular Weight (g/mol): 190.21
Molecular Formula: C7H10O4S
InChi Key: FHYQDMHVFCUXLL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)S(=O)(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results