missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Oxovalerate 98.0+%, TCI America™

Catalog No. K003525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K003525G 25 g
K0035100G 100 g
K0035500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K003525G Supplier TCI America Supplier No. K003525G

Chemical Identifiers

CAS 30414-53-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00011705
InChI Key XJMIXEAZMCTAGH-UHFFFAOYSA-N
Synonym methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
PubChem CID 121699
IUPAC Name methyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OC

Specifications

Color Yellow
Boiling Point 74°C
Quantity 25 g
Formula Weight 130.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Oxovalerate
EINECSNumber (2)-1505
TSCA No