missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Oxohexanoate 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 85°C |
| Quantity | 500 mL |
| Formula Weight | 144.17 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 3-Oxohexanoate |
Safety and Handling
| EINECSNumber | (2)-1505 |
| TSCA | No |