missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH6211006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6211006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6211006 Supplier Thermo Scientific Chemicals Supplier No. H6211006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Methyl 3-oxocyclobutanecarboxylate can be used as an intermediate for the synthesis of various pharmaceutical compounds. It is used for the preparation of novel imidazobenzazepine derivatives as dual H1/5-HT2A antagonists for the treatment of sleep disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-oxocyclobutanecarboxylate can be used as an intermediate for the synthesis of various pharmaceutical compounds. It is used for the preparation of novel imidazobenzazepine derivatives as dual H1/5-HT2A antagonists for the treatment of sleep disorders.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 695-95-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00604222
InChI Key IHLHSAIBOSSHQV-UHFFFAOYSA-N
Synonym methyl 3-oxocyclobutanecarboxylate, methyl 3-oxo-cyclobutanecarboxylate, 3-methoxycarbonylcyclobutanone, cyclobutanecarboxylic acid, 3-oxo-, methyl ester, 3-oxocyclobutanecarboxylic acid methyl ester, ksc630k4p, methyl 3-oxocyclobutane carboxylate, 3-oxo-1-cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylicacid, 3-oxo-, methyl ester
PubChem CID 13992367
IUPAC Name methyl 3-oxocyclobutane-1-carboxylate
SMILES COC(=O)C1CC(=O)C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 128.12
Percent Purity 97%
Chemical Name or Material Methyl 3-oxocyclobutanecarboxylate
Hazard Category H226-H317-H319
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P303+P361+P353-P305+P351+P338-P333+P313-P363-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only