CAS RN 695-95-4
IUPAC Name:
methyl 3-oxocyclobutane-1-carboxylate
Synonyms:
methyl 3-oxocyclobutane-1-carboxylate
Molecular Weight (g/mol):
128.13
Molecular Formula:
C6H8O3
InChi Key:
IHLHSAIBOSSHQV-UHFFFAOYSA-N
SMILES:
COC(=O)C1CC(=O)C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
Methyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals
CAS: 695-95-4 Molecular Formula: C6H8O3 Molecular Weight (g/mol): 128.13 MDL Number: MFCD00604222 InChI Key: IHLHSAIBOSSHQV-UHFFFAOYSA-N Synonym: methyl 3-oxocyclobutanecarboxylate,methyl 3-oxo-cyclobutanecarboxylate,3-methoxycarbonylcyclobutanone,cyclobutanecarboxylic acid, 3-oxo-, methyl ester,3-oxocyclobutanecarboxylic acid methyl ester,ksc630k4p,methyl 3-oxocyclobutane carboxylate,3-oxo-1-cyclobutanecarboxylic acid methyl ester,cyclobutanecarboxylicacid, 3-oxo-, methyl ester PubChem CID: 13992367 IUPAC Name: methyl 3-oxocyclobutane-1-carboxylate SMILES: COC(=O)C1CC(=O)C1