missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133652
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015725G 25 g
N0157500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N015725G Supplier TCI America Supplier No. N015725G

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007250
InChI Key AXLYJLKKPUICKV-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
PubChem CID 69260
IUPAC Name methyl 3-nitrobenzoate
SMILES COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 279°C
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Nitrobenzoate
EINECSNumber (3)-1503
TSCA Yes