missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-(Methylthio)propionate, TCI America™
Supplier: TCI America M086225G
Specifications
| Methyl 3-(Methylthio)propionate | |
| Colorless | |
| C5H10O2S | |
| 25 g | |
| DMMJVMYCBULSIS-UHFFFAOYSA-N | |
| methyl 3-(methylsulfanyl)propanoate | |
| 61641 | |
| Liquid |
| 13532-18-8 | |
| 81°C | |
| MFCD00008565 | |
| methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate | |
| COC(=O)CCSC | |
| 134.19 | |
| 134.19 |
Chemical Identifiers
| 13532-18-8 | |
| 134.19 | |
| DMMJVMYCBULSIS-UHFFFAOYSA-N | |
| 61641 | |
| COC(=O)CCSC |
| C5H10O2S | |
| MFCD00008565 | |
| methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate | |
| methyl 3-(methylsulfanyl)propanoate |
Safety and Handling
EINECSNumber : (2)-1395
TSCA : Yes