missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methylpyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. M27461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27461G 1 g
M27465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27461G Supplier TCI America Supplier No. M27461G
Only null left

Chemical Identifiers

CAS 59718-84-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD03789583
InChI Key CCQFKEITVOTHIW-UHFFFAOYSA-N
Synonym 3-Methylpyridine-2-carboxylic Acid Methyl Ester, Methyl 3-Methylpicolinate, 3-Methylpicolinic Acid Methyl Ester
PubChem CID 12264487
IUPAC Name methyl 3-methylpyridine-2-carboxylate
SMILES COC(=O)C1=C(C)C=CC=N1

Specifications

Color Colorless
Boiling Point 73°C
Quantity 1 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Methylpyridine-2-carboxylate