CAS RN 59718-84-2
CAS RN 59718-84-2
IUPAC Name:
methyl 3-methylpyridine-2-carboxylate
Synonyms:
methyl 3-methylpyridine-2-carboxylate
Molecular Weight (g/mol):
151.17
Molecular Formula:
C8H9NO2
InChi Key:
CCQFKEITVOTHIW-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(C)C=CC=N1
1
–
1
of
1
results
1
Methyl 3-Methylpyridine-2-carboxylate 98.0+%, TCI America™
CAS: 59718-84-2 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.17 MDL Number: MFCD03789583 InChI Key: CCQFKEITVOTHIW-UHFFFAOYSA-N Synonym: 3-Methylpyridine-2-carboxylic Acid Methyl Ester, Methyl 3-Methylpicolinate, 3-Methylpicolinic Acid Methyl Ester PubChem CID: 12264487 IUPAC Name: methyl 3-methylpyridine-2-carboxylate SMILES: COC(=O)C1=C(C)C=CC=N1