missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methylaminocrotonate, 97%

Catalog No. AAB2019914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2019914 25 g
AAB2019906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2019914 Supplier Thermo Scientific Chemicals Supplier No. B2019914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13412-12-9 | C6H11NO2 | 129.159 g/mol

It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.

Solubility
Soluble in water at (54 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13412-12-9
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00027383
InChI Key QAKYFFYZPIPLDN-SNAWJCMRSA-N
Synonym methyl 3-methylaminocrotonate, methyl e-3-methylamino but-2-enoate, methyl 2e-3-methylamino but-2-enoate, methyl 3-methylaminobut-2-enoate, beta-n-methylaminocrotonic acid methyl ester, methyl 3-methyl-amino crotonate, e-methyl 3-methylamino but-2-enoate, 3-methylamino isocrotonic acid methyl ester, 2-butenoic acid,3-methylamino-,methyl ester, e-3-methylamino-2-butenoic acid methyl ester
PubChem CID 5846045
IUPAC Name methyl (E)-3-(methylamino)but-2-enoate
SMILES CC(=CC(=O)OC)NC

Specifications

Melting Point 60°C to 63°C
Quantity 25 g
Solubility Information Soluble in water at (54g/L) (25°C),Calc.
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material Methyl 3-methylaminocrotonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only