CAS RN 13412-12-9
IUPAC Name:
methyl 3-(methylimino)butanoate
Synonyms:
methyl 3-(methylimino)butanoate
Molecular Weight (g/mol):
129.16
Molecular Formula:
C6H11NO2
InChi Key:
UPQBRLACXPZFPJ-UHFFFAOYSA-N
SMILES:
COC(=O)CC(C)=NC
1
–
1
of
1
results
1
Promotions Available
Methyl 3-methylaminocrotonate, 97%
Methyl 3-methylaminocrotonate, 97%
CAS: 13412-12-9 Molecular Formula: C6H11NO2 Molecular Weight (g/mol): 129.159 MDL Number: MFCD00027383 InChI Key: QAKYFFYZPIPLDN-SNAWJCMRSA-N Synonym: methyl 3-methylaminocrotonate,methyl e-3-methylamino but-2-enoate,methyl 2e-3-methylamino but-2-enoate,methyl 3-methylaminobut-2-enoate,beta-n-methylaminocrotonic acid methyl ester,methyl 3-methyl-amino crotonate,e-methyl 3-methylamino but-2-enoate,3-methylamino isocrotonic acid methyl ester,2-butenoic acid,3-methylamino-,methyl ester,e-3-methylamino-2-butenoic acid methyl ester PubChem CID: 5846045 IUPAC Name: methyl (E)-3-(methylamino)but-2-enoate SMILES: CC(=CC(=O)OC)NC