missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methyl-4-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133051
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21455G 5 g
M214525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21455G Supplier TCI America Supplier No. M21455G
Only null left

Chemical Identifiers

CAS 24078-21-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00085640
InChI Key IEFONJKJLZFGKQ-UHFFFAOYSA-N
Synonym benzoic acid, 3-methyl-4-nitro-, methyl ester, 3-methyl-4-nitrobenzoic acid methyl ester, m-toluic acid, 4-nitro-, methyl ester, methyl 3-methyl-4-nitrobenzenecarboxylate, benzoic acid, 3-methyl-4-nitro, methyl ester, methyl3-methyl-4-nitrobenzoate, pubchem2350, acmc-1cfdu, methyl 4-nitro-m-toluate, ksc205m3d
PubChem CID 260927
IUPAC Name methyl 3-methyl-4-nitrobenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]

Specifications

Melting Point 81°C
Color Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methyl-4-nitrobenzoate