missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methyl-4-nitrobenzoate, 97%

Catalog No. AAB2062706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2062706 5 g
AAB2062714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2062706 Supplier Thermo Scientific Chemicals Supplier No. B2062706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24078-21-5 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24078-21-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00085640
InChI Key IEFONJKJLZFGKQ-UHFFFAOYSA-N
Synonym benzoic acid, 3-methyl-4-nitro-, methyl ester, 3-methyl-4-nitrobenzoic acid methyl ester, m-toluic acid, 4-nitro-, methyl ester, methyl 3-methyl-4-nitrobenzenecarboxylate, benzoic acid, 3-methyl-4-nitro, methyl ester, methyl3-methyl-4-nitrobenzoate, pubchem2350, acmc-1cfdu, methyl 4-nitro-m-toluate, ksc205m3d
PubChem CID 260927
IUPAC Name methyl 3-methyl-4-nitrobenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)[N+](=O)[O-]

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Formula Weight 195.18
Percent Purity 97%
Chemical Name or Material Methyl 3-methyl-4-nitrobenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only