missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methyl-2-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133094
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22135G 5 g
M221325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22135G Supplier TCI America Supplier No. M22135G

Chemical Identifiers

CAS 5471-82-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD01003947
InChI Key NJHDBIXFFZVJGZ-UHFFFAOYSA-N
Synonym methyl3-methyl-2-nitrobenzoate, methyl 3-methyl-2-nitrobenzenecarboxylate, benzoic acid, 3-methyl-2-nitro-, methyl ester, 2-nitro-3-methylbenzoic acid, methyl ester, 3-methyl-2-nitrobenzoic acid methyl ester, pubchem10932, acmc-1ap0b, ksc495q9t, methyl 2-nitro-3-methylbenzoate, methyl 3-methyl-2-nitro-benzoate
PubChem CID 79621
IUPAC Name methyl 3-methyl-2-nitrobenzoate
SMILES CC1=CC=CC(=C1[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methyl-2-nitrobenzoate