missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methyl-2-nitrobenzoate, 98%

Catalog No. p-7054042
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6190714 25 g
AAH6190722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6190714 Supplier Thermo Scientific Chemicals Supplier No. H6190714
Only null left

CAS: 5471-82-9 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5471-82-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD01003947
InChI Key NJHDBIXFFZVJGZ-UHFFFAOYSA-N
Synonym methyl3-methyl-2-nitrobenzoate, methyl 3-methyl-2-nitrobenzenecarboxylate, benzoic acid, 3-methyl-2-nitro-, methyl ester, 2-nitro-3-methylbenzoic acid, methyl ester, 3-methyl-2-nitrobenzoic acid methyl ester, pubchem10932, acmc-1ap0b, ksc495q9t, methyl 2-nitro-3-methylbenzoate, methyl 3-methyl-2-nitro-benzoate
PubChem CID 79621
IUPAC Name methyl 3-methyl-2-nitrobenzoate
SMILES CC1=CC=CC(=C1[N+](=O)[O-])C(=O)OC

Specifications

Quantity 25 g
Formula Weight 195.17
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 3-methyl-2-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only