missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxythiophene-2-carboxylate, 97%

Catalog No. p-7025211
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2058603 1 g
AAB2058606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2058603 Supplier Thermo Scientific Chemicals Supplier No. B2058603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62353-75-7 | C7H8O3S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62353-75-7
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00102115
InChI Key MWADWXJPBMFRGL-UHFFFAOYSA-N
Synonym 3-methoxy-2-thiophenecarboxylic acid methyl ester, acmc-20abea, methyl 3-methoxythiophene carboxylate, methyl 3-methoxy-2-thiophenecarboxylate, 2-thiophenecarboxylicacid,3-methoxy-,methyl ester, 3-methoxythiophene-2-carboxylic acid methyl ester
PubChem CID 701637
IUPAC Name methyl 3-methoxythiophene-2-carboxylate
SMILES COC(=O)C1=C(OC)C=CS1

Specifications

Melting Point 60°C to 63°C
Quantity 1 g
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material Methyl 3-methoxythiophene-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only