missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxyphenylacetate, 97%

Catalog No. p-7024961
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2023788 2.5 g
AAB2023709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2023788 Supplier Thermo Scientific Chemicals Supplier No. B2023788

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18927-05-4 | C10H12O3 | 180.203 g/mol

Methyl 3-methoxyphenylacetate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methoxyphenylacetate is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 18927-05-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00017205
InChI Key BSVIOYCZTJRBDB-UHFFFAOYSA-N
Synonym methyl 2-3-methoxyphenyl acetate, methyl 3-methoxyphenylacetate, benzeneacetic acid, 3-methoxy-, methyl ester, acetic acid, m-methoxyphenyl-, methyl ester, methyl 3-methoxyphenyl acetate, 3-methoxy-phenyl-acetic acid methyl ester, acmc-1brer, methyl m-methoxyphenylacetate, methyl 3-methoxy-phenylacetate
PubChem CID 519609
IUPAC Name methyl 2-(3-methoxyphenyl)acetate
SMILES COC1=CC=CC(=C1)CC(=O)OC

Specifications

Boiling Point 86°C to 88°C (0.5 mmHg)
Quantity 2.5 g
Solubility Information Sparingly soluble in water.
Formula Weight 180.23
Percent Purity 97%
Chemical Name or Material Methyl 3-methoxyphenylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only