missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methoxy-4-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7132725
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14275G 5 g
M142725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14275G Supplier TCI America Supplier No. M14275G

Chemical Identifiers

CAS 3556-83-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00082710
InChI Key LLEXCSBUSVRBCA-UHFFFAOYSA-N
Synonym 3-methoxy-4-methylbenzoic acid methyl ester, methyl 3-methoxy-p-toluate, 4-methyl-m-anisic acid methyl ester, benzoic acid, 3-methoxy-4-methyl-, methyl ester, pubchem13650, methyl 4-methyl-m-anisate, acmc-1cs22, methyl3-methoxy-4-methylbenzoate, methyl 3-methoxy-4-methyl-benzoate, 3-methoxy-p-toluic acid methyl ester
PubChem CID 591123
IUPAC Name methyl 3-methoxy-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)OC

Specifications

Color White
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methoxy-4-methylbenzoate