missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxy-4-methylbenzoate, 98%

Catalog No. p-7026611
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2269506 5 g
AAB2269514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2269506 Supplier Thermo Scientific Chemicals Supplier No. B2269506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3556-83-0 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3556-83-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00082710
InChI Key LLEXCSBUSVRBCA-UHFFFAOYSA-N
Synonym 3-methoxy-4-methylbenzoic acid methyl ester, methyl 3-methoxy-p-toluate, 4-methyl-m-anisic acid methyl ester, benzoic acid, 3-methoxy-4-methyl-, methyl ester, pubchem13650, methyl 4-methyl-m-anisate, acmc-1cs22, methyl3-methoxy-4-methylbenzoate, methyl 3-methoxy-4-methyl-benzoate, 3-methoxy-p-toluic acid methyl ester
PubChem CID 591123
IUPAC Name methyl 3-methoxy-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)OC

Specifications

Melting Point 47°C to 50°C
Boiling Point 119°C to 120°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5295
Quantity 5 g
Beilstein 1948339
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxy-4-methylbenzoate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only