missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methoxy-2-naphthoate 98.0+%, TCI America™

Catalog No. M14625G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14625G Supplier TCI America Supplier No. M14625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13041-60-6
Molecular Formula C13H12O3
Molecular Weight (g/mol) 216.236
MDL Number MFCD00454253
InChI Key MPLSAOVNAXJHHI-UHFFFAOYSA-N
Synonym 3-Methoxy-2-naphthoic Acid Methyl Ester
PubChem CID 296952
IUPAC Name methyl 3-methoxynaphthalene-2-carboxylate
SMILES COC1=CC2=CC=CC=C2C=C1C(=O)OC

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 217°C
Quantity 5 g
Formula Weight 216.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methoxy-2-naphthoate