missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-iodo-4-methoxybenzoate, 98%

Catalog No. p-7080829
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1054906 5 g
AAA1054914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1054906 Supplier Thermo Scientific Chemicals Supplier No. A1054906
Only null left

CAS: 35387-93-0 | C9H9IO3 | 292.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35387-93-0
Molecular Formula C9H9IO3
Molecular Weight (g/mol) 292.07
MDL Number MFCD00052925
InChI Key GHNGBFHLUOJHKP-UHFFFAOYSA-N
Synonym 3-iodo-4-methoxy-benzoic acid methyl ester, 3-iodo-4-methoxybenzoic acid methyl ester, benzoic acid, 3-iodo-4-methoxy-, methyl ester, methyl3-iodo-4-methoxybenzoate, methyl 3-iodo-p-anisate, acmc-1agb4, ksc495q3h, methyl-3-iodo-4-methoxybenzoate, methyl 3-iodo-4-methoxy-benzoate, 3-iodo-p-anisic acid methyl ester
PubChem CID 316980
IUPAC Name methyl 3-iodo-4-methoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C(I)=C1

Specifications

Melting Point 93°C to 95°C
Quantity 5 g
Beilstein 2692711
Sensitivity Light sensitive
Formula Weight 292.07
Percent Purity 98%
Chemical Name or Material Methyl 3-iodo-4-methoxybenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only