missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-Hydroxy-4-nitrobenzoate 98.0+%, TCI America™
$132.35 - $441.78
Chemical Identifiers
| CAS | 713-52-0 |
|---|---|
| Molecular Formula | C8H7NO5 |
| Molecular Weight (g/mol) | 197.146 |
| MDL Number | MFCD00016997 |
| InChI Key | UEGCRFNWTGYVKX-UHFFFAOYSA-N |
| Synonym | benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate |
| PubChem CID | 294866 |
| IUPAC Name | methyl 3-hydroxy-4-nitrobenzoate |
| SMILES | COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O |
Chemical Identifiers
| 713-52-0 | |
| 197.146 | |
| UEGCRFNWTGYVKX-UHFFFAOYSA-N | |
| 294866 | |
| COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O |
| C8H7NO5 | |
| MFCD00016997 | |
| benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate | |
| methyl 3-hydroxy-4-nitrobenzoate |
Specifications
| 713-52-0 | |
| Yellow | |
| MFCD00016997 | |
| benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate | |
| COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O | |
| 197.146 | |
| 197.15 | |
| Crystalline Powder |
| 92°C | |
| C8H7NO5 | |
| 5 g | |
| UEGCRFNWTGYVKX-UHFFFAOYSA-N | |
| methyl 3-hydroxy-4-nitrobenzoate | |
| 294866 | |
| ≥98.0% (GC) | |
| Methyl 3-Hydroxy-4-nitrobenzoate |
Safety and Handling
TSCA : No