missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxy-4-nitrobenzoate 98.0+%, TCI America™

Catalog No. M22505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22505G 5 g
M225025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22505G Supplier TCI America Supplier No. M22505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 713-52-0
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00016997
InChI Key UEGCRFNWTGYVKX-UHFFFAOYSA-N
Synonym benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate
PubChem CID 294866
IUPAC Name methyl 3-hydroxy-4-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 92°C
Color Yellow
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Hydroxy-4-nitrobenzoate